Learn More
Simport™ Scientific Histosette™ II Tissue Processing/Embedding Cassette in Quickload™ Stacks
Secure specimens during processing with Simport Scientific™ Histosette™ II Tissue Processing/Embedding Cassette. Keep specimens safely submerged in liquid. The flow-through slots maximize fluid exchange. The one-piece integral lid eliminates the need for separate steel lids.
Specifica
Materiale Acetal
Quantità 2000
Visualizza altre specifiche
Descrizione
Descrizione
In Quickload™ sleeve
  • 45° anterior writing area
  • Open and close as often as necessary — cassettes will always relock securely
  • Back mounted tab allowing an unobstructed anterior marking area
  • Suitable for automated printers
  • Made of acetal
  • Resistant to the chemical action of most histological solvents
Order Info
Each case contains 50 Quickload™ stacks of 40 cassettes.
Certifications
CE-IVD
Specifica
Specifica
Acetal
2000