Learn More
MP Biomedicals HIV Blot 2.2 (CE)
Sfoglia Tutto MP Biomedicals Prodotti
Click to view available options
Quantità:
18 Tests/Kit
36 Tests/Kit
Dimensione della confezione:
1 set
Descrizione
- The nitrocellulose strips are precoated with antigenic proteins from partially purified inactivated HIV-1, and a specific HIV-2 synthetic peptide on the same strips.
- Individual nitrocellulose strips are incubated with diluted serum or plasma and controls.
- Specific antibodies to HIV-1 and HIV-2, if present in the specimens, will bind to the HIV-1 proteins and HIV-2 peptide on the strips.
- The strips are washed to remove unbound materials.
- Antibodies that bind specifically to HIV proteins can be visualized using a series of reactions with goat anti-human IgG conjugated with alkaline phosphatase and the substrate BCIP/NBT.
- Each strip also includes an internal sample addition control to minimize the risk of false negatives due to operational errors and to ensure the addition of samples.
- CE-marked.
- For professional use only.
- Human serum and plasma.
Specifica
Specifica
| Product Type | Western Blot Kit |
| Metodo di rilevamento | Colorimetric |
| Da utilizzare con (applicazione) | Immunoblot (Western) |
| Membrane Compatibility | Nitrocellulose |
| Quantità | 18 Tests/Kit |
| Substrate | Alkaline Phosphatase |