Learn More
MP Biomedicals HIV Blot 2.2 (RUO)
Codice prodotto. p-200136116 Sfoglia Tutto MP Biomedicals Prodotti
Click to view available options
Quantità:
18 Tests/Kit
36 Tests/Kit
Dimensione della confezione:
1 set
Codice prodotto. 30694936
Marca: MP Biomedicals 11032018
The Blot is a qualitative enzyme immunoassay for in vitro detection of antibodies to HIV-1 and HIV-2 in human serum or plasma.
  • The nitrocellulose strips are precoated with antigenic proteins from partially purified inactivated HIV-1, and a specific HIV-2 synthetic peptide on the same strips.
  • Individual nitrocellulose strips are incubated with diluted serum or plasma and controls. Specific antibodies to HIV-1 and HIV-2, if present in the specimens, will bind to the HIV-1 proteins and HIV-2 peptide on the strips.
  • The strips are washed to remove unbound materials.
  • Antibodies that bind specifically to HIV proteins can be visualized using a series of reactions with goat anti-human IgG conjugated with alkaline phosphatase and the substrate BCIP/NBT.
  • Each strip also includes an internal sample addition control to minimize the risk of false negatives due to operational errors and to ensure the addition of samples.
  • It is intended for use as a more specific supplemental test on human serum or plasma specimens found repeatedly reactive using screening procedures such as ELISA.
  • Research Use Only (RUO).
  • For professional use only.
  • Human serum and plasma.
Specifica
Product Type Western Blot Kit
Metodo di rilevamento Colorimetric
Da utilizzare con (applicazione) Immunoblot (Western)
Membrane Compatibility Nitrocellulose
Quantità 18 Tests/Kit
Substrate Alkaline Phosphatase