Learn More
IBA Lifesciences™ HIV p24-Gag Peptide KAFSPEVIPMF (HLA-B*5701)
Sfoglia Tutto IBA Lifesciences Prodotti
Click to view available options
Quantità:
1 mg
Dimensione della confezione:
1mg
Descrizione
HIV p24-Gag peptide KAFSPEVIPMF (HLA-B*5701) is a single peptide for stimulation of T cells. The peptide from human immunodeficiency virus 1 (HIV-1) p24 group-specific antigen (Gag) is synthesized as it is presented by the MHC class I HLA-B*5701 allele and can be used for targeted in vitro generation and expansion of antigen-specific CD8+ T cells.
- Peptides have been purified by HPLC using a water and acetonitrile gradient
- Endotoxin-free
Specifica
Specifica
| Purezza | >85% |
| Livello di endotossine | Endotoxin-free |
| Da utilizzare con (applicazione) | T Cell Stimulation |
| Liofilizzato | Yes |
| Quantità | 1 mg |