missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HLA-F Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 14670-1-AP-150UL
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
HLA-F belongs to the HLA class I heavy chain paralogues. It is a non-classical heavy chain that forms a heterodimer with a beta-2 microglobulin light chain, with the heavy chain anchored in the membrane. Unlike most other HLA heavy chains, this molecule is localized in the endoplasmic reticulum and Golgi apparatus, with a small amount present at the cell surface in some cell types. It contains a divergent peptide-binding groove, and is thought to bind a restricted subset of peptides for immune presentation.Specifica
| HLA-F | |
| Polyclonal | |
| Unconjugated | |
| HLA-F | |
| CDA12, HLA 5.4, HLA CDA12, HLA F, HLA F antigen, HLAF, HLA-F, Leukocyte antigen F, MHC class I antigen F | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 3134 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P30511 | |
| HLA-F | |
| HLA-F Fusion Protein Ag6335 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto