Learn More
Epredia™ HM 525 NX Cryostat Blade Holders
Sfoglia Tutto Epredia Prodotti
Click to view available options
Product Type:
Disposable Blade Carrier EC
Disposable Blade Carrier EC70
Standard Knife Holder
Da utilizzare con (apparecchiatura):
HM 525 NX and HM 550
Dimensione della confezione:
Pezzo
Descrizione
Standard Knife Carrier:
- Allows for insertion of standard knives, with clearance angle adjustment
- Can be moved horizontally on precision dovetail guide with quick clamp system
- Includes anti-roll plate
Disposable Blade Carrier EC:
- Allows for insertion of high- and low-profile blades, with clearance angle adjustment
- Can be moved on precision dovetail guide with quick clamp plate
- Includes anti-roll plate
Disposable Blade Carrier EV:
- Use with vacutome system
- Allows for insertion of high- and low-profile disposable blades
- Includes a vacuum anti-roll hood for stretching and disposing of section waste
- Features a slot that is opened and closed by a vacuum-controlled flap
Magnetic Disposable Blade Carrier MV:
- Use with vacutome system
- Allows for insertion of disposable blades
- Includes a vacuum anti-roll hood for stretching and disposing of section waste
- Features a slot that is opened and closed by a vacuum-controlled flap
Specifica
Specifica
| Product Type | Disposable Blade Carrier EC70 |
| Da utilizzare con (apparecchiatura) | HM 525 NX and HM 550 |