missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HMBS Polyclonal Antibody
Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA579372
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: human Hela whole cell, human placenta tissue, human HepG2 whole cell, human 22RV1 whole cell, human U20S whole cell.
PBGD (porphobilinogen deaminase), also designated hydroxymethylbilane synthase, is a cytoplasmic enzyme found in the heme synthesis pathway. PBGD belongs to the HMBS (hydroxymethylbilane synthase) family. Deficiency of PBGD causes errors in pyrrole metabolism, which in turn leads to an inherited autosomal disorder called acute intermittent porphyria (AIP). AIP is characterized by acute attacks of neurological dysfunctions with hypertension, tachycardia, peripheral neurologic disturbances, abdominal pain and excessive amounts of aminolevulinic acid and porphobilinogen in the urine.
Specifica
| HMBS | |
| Polyclonal | |
| Unconjugated | |
| Hmbs | |
| alternative name: porphobilinogen deaminase; HEM3; hemC; HMBS; Hydroxymethylbilane synthase; PBG D; PBGD; PBG-D; PORC; Porphobilinogen deaminase; porphyria, acute; Chester type; pre-uroporphyrinogen synthase; similar to Porphobilinogen deaminase (Hydroxymethylbilane synthase) (HMBS) (Pre-uroporphyrinogen synthase) (PBG-D); T25658; Ups; uroporphyrinogen I synthase; uroporphyrinogen I synthetase; URO-S; Uros1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 15288, 25709, 3145 | |
| -20°C | |
| Lyophilized |
| ELISA, Western Blot | |
| 500 μg/mL | |
| PBS with 4mg trehalose and 0.05mg sodium azide | |
| P08397, P19356, P22907 | |
| Hmbs | |
| E. coli-derived human HMBS recombinant protein (Position: N6-H361). | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Correzione del contenuto del prodotto
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion