Learn More
HO-1/HMOX1 Mouse anti-Human, CoraLite 488, Clone: 2D10A5, Proteintech
Mouse Monoclonal Antibody
Marca: Proteintech CL488-66743-100UL
Descrizione
The HO-1/HMOX1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human HO-1/HMOX1. This antibody recognizes human, mouse, rat, pig antigen. The HO-1/HMOX1 antibody has been validated for the following applications: FC (Intra), IF analysis. This antibody is coralite™488.
Specifica
| HO-1/HMOX1 | |
| Monoclonal | |
| CoraLite 488 | |
| 100 μL | |
| 3162 | |
| IgG2a |
| Immunofluorescence, Flow Cytometry (Intracellular Staining) | |
| 2D10A5 | |
| Mouse | |
| Primary | |
| Human, Mouse, Rat, Pig |