missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Hoechst 33258 analog 2

Codice prodotto. 16472777
Encompass_Preferred
Click to view available options
Quantità:
5 mg
10 mg
50 mg
100 mg
Dimensione della confezione:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16472777

Brand: MedChemExpress HY15624

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!

This item is not returnable. View return policy

MedChemExpress Hoechst 33258 analog 2 are part of a family of blue fluorescent dyes used to stain DNA.

Specifications

Content And Storage 4°C, protect from light ∣ In solvent : -80°C, 6 months ∣ -20°C, 1 month (protect from light)
Quantità 50 mg
Shipping Condition Room temperature
Solubilità H2O : < 0.1 mg/mL (insoluble)
Peso molecolare 422.52 g/mol
CAS 23491-54-5
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.