Learn More
Hoefer™ Gel Seal
Codice prodotto. 11955338
Click to view available options
:
Gel seal; 0.25 oz.
Codice prodotto. 11955338
Marca: Hoefer™ SE6070
A sealant used very sparingly in vertical gel casting to reduce leaking
  • For use with E245 Dual Gel Caster
Specifica
Tipo Gel Seal
Volume (sistema imperiale) 0.25 oz.