Learn More
Kinematica™ Polytron™ PT 10-35 GT Homogenzier Drive Unit
Codice prodotto. 10103703
Cambia vista
Click to view available options
Tensione:
230 V
Dimensione della confezione:
Pezzo
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Tensione unitSize
10103703 230 V Pezzo
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Codice prodotto. 10103703 Fornitore Kinematica™ N. del fornitore 11010104
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
Compact,economical homogenizer with 1200 W motor for processing larger samples. Versatile design allows for samples as small as .1 ml and up to 4500 ml with appropriate dispersing aggregate. Over 25 aggregates are available for almost any dispersing or homogenizing application.
  • Application volumes 0.1mL to 10L
  • 1200W motor provides sufficient power and high speed necessary for efficient particle size reduction
  • Digital motor speed control from 500 to 30,000 rpm
  • Aggregate tip speeds up to 32 m/s produce the finest particles possible
  • Soft start prevents splashing of homogenate from sample vessel
Specifica
Product Type Homogenizer Drive Unit
Tensione 230 V
Intervallo volume (sistema metrico) 0.1 mL to 10 L
Da utilizzare con (applicazione) Difficult-to-process Samples