missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HPV16 E1+E4 Antibody (HPV16 E1/E4), DyLight 350, Novus Biologicals™
Mouse Monoclonal Antibody
Marca: Novus Biologicals NBP3-08319UV
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
HPV16 E1+E4 Monoclonal antibody specifically detects HPV16 E1+E4 in Virus samples. It is validated for Immunohistochemistry (Paraffin)
Specifica
| HPV16 E1+E4 | |
| Monoclonal | |
| DyLight 350 | |
| ATP dependent helicase E1, E1, E4, HPV 16 ATP dependent helicase E1, HPV 16 E1, HPV 16 E4, HPV 16 replication protein E1, HPV16 ATP dependent helicase E1, HPV16 E1, HPV16 E4, HPV16 replication protein E1, Human papilloma virus type 16, Human papilloma virus type 16 ATP dependent helicase E1, Human papilloma virus type 16 E1, Human papilloma virus type 16 E4, Human papilloma virus type 16 replication protein E1, Human papillomavirus type 16, Human papillomavirus type 16 ATP dependent helicase E1, Human papillomavirus type 16 E1, Human papillomavirus type 16 E4, Human papillomavirus type 16 replication protein E1, Replication protein, Replication protein E1 | |
| Recombinant human HPV16 E1+E4 fragment (aa 36-41). | |
| 100 μg | |
| Cancer, Virology Bacteria and Parasites | |
| Virus | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| HPV16 E1/E4 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG1 κ |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto