Learn More
Horst™ HSS Model Flexible Heating Conductor
missing translation for 'orderingAttributeHoverText'
Dimensioni (L x P):
0.6 m
10 m
1 m
2 m
3 m
4 m
5 m
6 m
missing translation for 'unitSize'
Pezzo
Specifications
Specifications
| Dimensioni (L x P) | 1 m |
| Dimensioni (L x P x A) | 1 m |
| Requisiti elettrici | Tensione ∼230 V |
| Modello | HSS |
| Product Type | Conduttore riscaldante flessibile |