Learn More
HTR6 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 23561-1-AP-20UL
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
This is one of the several different receptors for 5-hydroxytryptamine, a biogenic hormone that functions as a neurotransmitter, a hormone, and a mitogen. The activity of this receptor is mediated by G proteins that stimulate adenylate cyclase. It has a high affinity for tricyclic psychotropic drugs.Specifica
| HTR6 | |
| Polyclonal | |
| Unconjugated | |
| HTR6 | |
| 5 HT 6, 5 HT6, 5 HT6R, 5 hydroxytryptamine receptor 6, HTR6, Serotonin receptor 6 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 3362 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.19 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P50406 | |
| HTR6 | |
| HTR6 Fusion Protein Ag15396 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |