Learn More
R&D Systems™ Human Angiopoietin-like 6 DuoSet ELISA
Click to view available options
Quantità:
1 Kit (for 5 Plates)
Dimensione della confezione:
pezzi 5
Descrizione
Includes
Capture antibody, detection antibody, recombinant standard, streptavidin conjugated to horseradish-peroxidase (Streptavidin-HRP) and development protocols
The Human Angiopoietin-like 6 DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies human Angiopoietin-like Protein 6/ANGPTL6.
Specifica
Specifica
| Quantità | 1 Kit (for 5 Plates) |
| Specie | Human |
| Coniugato | Biotin |
| Bersaglio legante | Angiopoietin-like Protein 6/ANGPTL6 |
| Da utilizzare con (applicazione) | ELISA |
| ID gene (immissione) | 83854 |
| Sinonimo | AGF, AGFAngiopoietin-related protein 5, Angiopoietin like Protein 6, angiopoietin-like 6, Angiopoietin-like protein 6, Angiopoietin-related growth factor, ANGPTL6, ARP5, ARP5angiopoietin-related protein 6 |
| Content And Storage | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |