Learn More
Human beta-Actin Antibody, R&D Systems™
Mouse Monoclonal Antibody has been used in 3 publications
Marca: R&D Systems MAB8969-SP
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
beta-Actin Monoclonal specifically detects beta-Actin in Human samples. It is validated for Immunocytochemistry.
Specifica
| beta-Actin | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 um filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 um filtered solution in PBS. with No Preservative | |
| ACTB, actin, beta, actin, cytoplasmic 1, B-Actin, Beta Actin, beta cytoskeletal actin, betaActin, Beta-actin, BRWS1, I(2)-Actin, PS1TP5-binding protein 1, PS1TP5BP1 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 60 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
| Immunocytochemistry | |
| 937210 | |
| Immunocytochemistry 8-25 ug/mL | |
| P60709 | |
| ACTB | |
| Peptide containing a sequence at the N-terminus of human beta-Actin Accession No. P60709 | |
| 25 ug | |
| Primary | |
| Detects human beta-Actin in direct ELISAs. | |
| Human | |
| Purified |