missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human BLIMP1/PRDM1 PE-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Marca: R&D Systems IC36081P
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
BLIMP1/PRDM1 Monoclonal specifically detects BLIMP1/PRDM1 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.
Specifica
| BLIMP1/PRDM1 | |
| Monoclonal | |
| R-PE | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| Beta-interferon gene positive regulatory domain I-binding factor, beta-interferon gene positive-regulatory domain I binding factor, BLIMP-1, BLIMP1MGC118925, blmp-1, B-lymphocyte-induced maturation protein 1, MGC118922, MGC118923, Positive regulatory domain I-binding factor 1, PR domain containing 1, with ZNF domain, PR domain zinc finger protein 1, PR domain-containing protein 1, PRDI-BF1MGC118924, PRDI-binding factor 1, PRDI-binding factor-1, PRDM1, PR-domain zinc finger protein 1, ZNFPR1A1 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 639 | |
| Human | |
| Purified |
| Flow Cytometry | |
| 646702 | |
| Intracellular Staining by Flow Cytometry 10 uL/10^6 cells | |
| O75626 | |
| PRDM1 | |
| E. coli-derived recombinant human BLIMP1/PRDM1 Lys667-Cys789 Accession # O75626 | |
| 100 Tests | |
| Primary | |
| Detects human BLIMP1/PRDM1 in Western blots. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
| IgG1 |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto