missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human c-Abl Antibody, R&D Systems™
Goat Polyclonal Antibody
Marca: R&D Systems AF5414
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
c-Abl Polyclonal specifically detects c-Abl in Human samples. It is validated for Western Blot, Simple Western.
Specifica
| c-Abl | |
| Polyclonal | |
| Unconjugated | |
| NP_009297 | |
| ABL1 | |
| E. coli-derived recombinant human c-Abl Ala941-Val1140 Accession # NP_009297 | |
| 100 μg | |
| Primary | |
| Detects human c-Abl in Western blots. | |
| Human | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 1 ug/mL, Simple Western 10 ug/mL | |
| Abelson murine leukemia viral oncogene homolog 1, ABL, ABL1, bcr/abl, BCR-ABL1, cAbl, c-ABL, c-abl oncogene 1, non-receptor tyrosine kinase, c-abl oncogene 1, receptor tyrosine kinase, EC 2.7.10, EC 2.7.10.2, JTK7, JTK7bcr/abl, p150bcr/c-abl oncogene protein, Proto-oncogene c-Abl, proto-oncogene tyrosine-protein kinase ABL1, tyrosine-protein kinase ABL1, v-abl, v-abl Abelson murine leukemia viral oncogene homolog 1 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 25 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto