missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human C-Reactive Protein/CRP Biotinylated Antibody, R&D Systems™
Sheep Polyclonal Antibody
Marca: R&D Systems BAF1707
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
C-Reactive Protein/CRP Polyclonal specifically detects C-Reactive Protein/CRP in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifica
| C-Reactive Protein/CRP | |
| Polyclonal | |
| Biotin | |
| P02741 | |
| CRP | |
| Mouse myeloma cell line NS0-derived recombinant human C-Reactive Protein/CRP Phe17-Pro224 Accession # P02741 | |
| 50 μg | |
| Primary | |
| Detects C-Reactive Protein/CRP in Western blots. In Western blots, approximately 20% cross-reactivity with recombinant mouse CRP is observed and 1% cross-reactivity with recombinant rat CRP is observed. | |
| Human | |
| IgG |
| Western Blot, Immunohistochemistry | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
| C-reactive protein, C-reactive protein, pentraxin-related, CRP, MGC88244, pentraxin 1, PTX1MGC149895 | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 1401 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto