missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Caveolin-1 Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 2 publications
Marca: R&D Systems AF5736
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Caveolin-1 Polyclonal specifically detects Caveolin-1 in Human samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Knockout Validated.
Specifica
| Caveolin-1 | |
| Polyclonal | |
| Unconjugated | |
| Q03135 | |
| CAV1 | |
| E. coli-derived recombinant human Caveolin-1 Ser2-Ser104 Accession # Q03135 | |
| 100 μg | |
| Primary | |
| Detects human Caveolin-1 in Western blots. | |
| Human | |
| IgG |
| Western Blot, Immunohistochemistry, KnockDown | |
| LYOPH | |
| Western Blot 0.2 ug/mL, Simple Western 2 ug/mL, Immunohistochemistry 5-15 ug/mL, Knockout Validated | |
| CAV, CAV1, caveolae protein, 22kD, caveolin 1, caveolae protein, 22kDa, Caveolin1, cell growth-inhibiting protein 32, MSTP085, VIP21 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 857 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto