Learn More
ACROBiosystems Human CD133 Full Length Protein, His Tag (Nanodisc)
Marca:  ACROBiosystems CD3-H52H1-20ug
Dettagli aggiuntivi : Peso : 0.00010kg
Codice prodotto. 17286090
  • 1826.84€ / 20µg
Spedizione stimata: 07-03-2024
per visualizzare lo stock disponibile
Descrizione
Descrizione
Prominin-1 is also known as CD133, Antigen AC133, PROM1, PROML1 and MSTP061. Is used as marker for hematopoietic stem and progenitor cells (HSPC) for somatic stem cell isolation. May play a role in cell differentiation, proliferation and apoptosis . Binds cholesterol in cholesterol-containing plasma membrane microdomains and may play a role in the organization of the apical plasma membrane in epithelial cells. During early retinal development acts as a key regulator of disk morphogenesis. Involved in regulation of MAPK and Akt signaling pathways. In neuroblastoma cells suppresses cell differentiation such as neurite outgrowth in a RET-dependent manner.
Nanodiscs are a new class of model membranes that are being used to solubilize and study a range of integral membrane proteins (mp, full length CD133) and membrane-associated proteins. The Nanodisc bilayer is bounded by a membrane scaffold protein (MSP1D1
Specifica
Specifica
NP_006008.1
20 µg
Gly 20 - His 865
CD133,PROM1,PROML1,Prominin-1,AC133
Unconjugated
His Tag
Less than 1.0 EU / µg by the LAL method.
Powder
>90% as determined by SDS-PAGE.
97.2 kDa
Human CD133 Full Length, His Tag (Nanodisc) (CD3-H52H1) is expressed from human 293 cells (HEK293). It contains AA Gly 20 - His 865 (Accession # O43490-1).
-20°C
CD133
Human
20 mM HEPES, 150 mM NaCl, pH7.5
HEK293
Bio-Markers & CD Antigens
Raccomandazioni
Raccomandazioni