missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CD42b/GPIb alpha Antibody, R&D Systems™

Mouse Monoclonal Antibody
Marca: R&D Systems MAB4067-SP
194.65 EUR valido fino al 2025-12-16
MIGLIOR PREZZO in promozione! Usa il codice promo "24090" per avere il tuo prezzo promozionale.
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
CD42b/GPIb alpha Monoclonal specifically detects CD42b/GPIb alpha in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.
Specifica
| CD42b/GPIb alpha | |
| Monoclonal | |
| LYOPH | |
| Western Blot 2 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready | |
| P07359 | |
| GP1BA | |
| Mouse myeloma cell line NS0-derived recombinant human CD42b/GPIb alpha His17-Leu505 Accession # P07359 | |
| 25 μg | |
| Primary | |
| Detects human CD42b/GPIb alpha in direct ELISAs and Western blots. | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry, CyTOF | |
| 486805 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Antigen CD42b-alpha, BP1BA, BSS, CD42b, CD42b antigen, CD42b-alpha, glycoprotein Ib (platelet), alpha polypeptide, Glycoprotein Ibalpha, GP1B, GP1BA, GPIb alpha, GP-Ib alpha, GPIbA, GPIb-alpha, MGC34595, platelet glycoprotein Ib alpha chain, platelet membrane glycoprotein 1b-alpha subunit | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 2811 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
For Research Use Only
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto