missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human CD44 (NP_000601) VersaClone cDNA
Click to view available options
Quantità:
10 μg
Dimensione della confezione:
10µg
Descrizione
- Also known as: IN; LHR; MC56; MDU2; MDU3; MIC4; Pgp1; CDW44; CSPG8; HCELL; HUTCH-I; ECMR-III
- CD44 is a cell-surface glycoprotein involved in cell-cell interactions, cell adhesion and migration.
- It is a receptor for hyaluronic acid (HA) and can also interact with other ligands, such as osteopontin, collagens, and matrix metalloproteinases (MMPs).
- It participates in a wide variety of cellular functions including lymphocyte activation, recirculation and homing, hematopoiesis, and tumor metastasis.
- Alternatively spliced transcripts encoding different proteins have been described.
- The insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
- For reasearch use only. Not for use in humans.
Specifica
Specifica
| Gene | hCD44 |
| Numero di accesso | NP_000601 |
| Concentrazione | 10 μg at 0.2 μg/μL |
| Formulazione | cDNA is provided in 10 mM Tris-Cl, pH 8.5 |
| Tipo di inserto | 2242 bp |
| ID gene (immissione) | 960 |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Specie | Human |
| Quantità | 10 μg |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto