Learn More
Invitrogen™ Human CD63 ELISA Kit
Codice prodotto. 16853020
Click to view available options
Quantità:
96 Tests
Dimensione della confezione:
96 tests
Codice prodotto. 16853020
Marca: Invitrogen™ EH95RB
ELISA
Human CD63 quantitates human CD63 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human CD63.
CD63 (LAMP-3, lysosome-associated membrane protein-3), a glycoprotein of tetraspanin family, is present in late endosomes, lysosomes and secretory vesicles of various cell types. CD63 is also present in the plasma membrane, usually following cell activation. Hence, CD63 has become a widely used basophil activation marker. In mast cells, however, CD63 exposition does not need their activation. CD63 interacts with integrins and affects phagocytosis and cell migration, it is also involved in H/K-ATPase trafficking regulation of ROMK1 channels. CD63 also serves as a T-cell costimulation molecule. Expression of CD63 can be used for predicting the prognosis in earlier stages of carcinomas. CD63 is expressed on activated platelets, and is a lysosomal membrane glycoprotein that is translocated to plasma membrane after platelet activation. CD63 is also present in monocytes and macrophages and is weakly expressed on granulocytes, B, and T cells. CD63 is identical to the melanoma-associated antigen which is ME491 and to the platelet antigen PTLGP40. Diseases associated with CD63 dysfunction include melanoma and Hermansky-Pudlak Syndrome.
TRUSTED_SUSTAINABILITY
Specifica
Numero di accesso C75951; CD 63; CD63; CD63 antigen; CD63 antigen (melanoma 1 antigen); Cd63 molecule; CD63 protein; granulophysin; LAMP-3; Lysosomal-associated membrane protein 3; lysosome-associated membrane glycoprotein 3; mast cell antigen AD1; ME491; melanoma 1 antigen; melanoma-associated antigen ME491; melanoma-associated antigen MLA1; MLA1; Ocular melanoma-associated antigen; OMA81H; tetraspanin-30; TSPAN30; Tspan-30
Intervallo saggio 14 to 3,000 pg/mL
Assay Sensitivity 14 pg/mL
Coniugato HRP
Product Type ELISA
Tipo di campione Plasma, Serum, Supernatant
Da utilizzare con (apparecchiatura) Colorimetric Microplate Reader
ID gene (immissione) 967
Simbolo del gene CD63
Interassay CV <12%
Intraassay CV <10
Contenuto kit Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantità 96 Tests
Status giuridico RUO
Volume del campione Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Requisiti di stoccaggio 2°C to 8°C
Bersaglio CD63
Durata del test 1 hrs 20 mins
Tempo totale saggio 4 hrs 45 mins