missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Cerberus 1 Antibody, R&D Systems™
Mouse Monoclonal Antibody
Marca: R&D Systems MAB1075
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Cerberus 1 Monoclonal specifically detects Cerberus 1 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifica
| Cerberus 1 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL | |
| O95813 | |
| CER1 | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Cerberus 1 Thr18-Ala267 (predicted) Accession # O95813 | |
| 500 μg | |
| Primary | |
| Detects human Cerberus 1 in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant human (rh) DAN, rhGremlin, or recombinant chicken Caronte is observed. | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry | |
| 162836 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CER1, cerberus, cerberus 1 (Xenopus laevis) homolog (cysteine knot superfamily), cerberus 1, cysteine knot superfamily, homolog (Xenopus laevis), cerberus-related 1, Cerberus-related protein, DAN domain family member 4, DAND4, MGC119894, MGC119895, MGC96951 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 9350 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto