missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Coronavirus Nucleocapsid Protein Rabbit anti-HCoV-OC43, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
214.00€ - 460.00€
Specifica
| Antigene | Human Coronavirus Nucleocapsid Protein |
|---|---|
| Applicazioni | Western Blot |
| Classificazione | Polyclonal |
| Coniugato | Unconjugated |
| Forma | Purified |
| Product Code | Brand | Quantità | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantità | Price | Quantity & Availability | |||||
|
18384556
|
Novus Biologicals
NBP3-16059-20UL |
20 μg |
214.00€
20µL |
Effettua il login per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso! | |||||
|
18354687
|
Novus Biologicals
NBP3-16059-100UL |
100 μg |
460.00€
100µL |
Effettua il login per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso! | |||||
Description
Human Coronavirus Nucleocapsid Protein Polyclonal antibody specifically detects Human Coronavirus Nucleocapsid Protein in HCoV-OC43 samples. It is validated for Western BlotSpecifications
| Human Coronavirus Nucleocapsid Protein | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS, 50% glycerol, pH7.3 | |
| IgG | |
| Affinity purified |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| HCoV-OC43 | |
| A synthetic peptide corresponding to a sequence within amino acids 100-200 of coronavirus Nucleoprotein (YP_009555245.1). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title