missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Creatine Kinase MM/CKMM Antibody, R&D Systems™

Mouse Monoclonal Antibody
Marca: R&D Systems MAB5564
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Creatine Kinase, Muscle/CKMM Monoclonal specifically detects Creatine Kinase, Muscle/CKMM in Human samples. It is validated for Western Blot, Simple Western.
Specifica
| Creatine Kinase, Muscle/CKMM | |
| Monoclonal | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Simple Western 1 ug/mL | |
| CKM, CKMM, CKMMcreatine kinase M-type, Creatine Kinase Isoenzyme, Creatine kinase M chain, creatine kinase, muscle, creatine kinase-M, EC 2.7.3, EC 2.7.3.2, M-CK | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 1158 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
| Western Blot | |
| 492731 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CKM | |
| Human cardiac muscle-derived Creatine Kinase, Muscle/CKMM | |
| 100 μg | |
| Primary | |
| Detects human Creatine Kinase, Muscle/CKMM in direct ELISAs and Western blots. | |
| Human | |
| Purified |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto