missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CTLA-4 (Research Grade Ipilimumab Biosimilar) Alexa Fluor 594-conjugated Antibody, R&D Systems™
Human Monoclonal Antibody
Marca: R&D Systems FAB9928T-100UG
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
CTLA-4 Monoclonal specifically detects CTLA-4 in Human samples. It is validated for Flow Cytometry.
Specifica
| CTLA-4 | |
| Monoclonal | |
| Alexa Fluor 594 | |
| Supplied 0.2 mg/ml in a saline solution containing BSA and Sodium Azide. | |
| Human | |
| Protein A or G purified from cell culture supernatant | |
| Primary | |
| Detects Human CTLA-4 based on Ipilimumab therapeutic antibody.This non-therapeutic antibody uses the same variable region sequence as the therapeutic antibody Ipilimumab. This product is for research use only. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
| IgG1 |
| Flow Cytometry | |
| Hu111 | |
| Flow Cytometry 0.25-1 μg/10^6 cells | |
| CD, CD152, CD152 antigen, CD152IDDM12, CD28, celiac disease 3, CELIAC3, CTLA4, CTLA-4, cytotoxic T-lymphocyte antigen 4, cytotoxic T-lymphocyte protein 4, Cytotoxic T-lymphocyte-associated antigen 4, cytotoxic T-lymphocyte-associated protein 4, cytotoxic T-lymphocyte-associated serine esterase-4, GRD4, GSE, ICOS, ligand and transmembrane spliced cytotoxic T lymphocyte associated antigen 4 | |
| Human CTLA-4 | |
| 100 μg | |
| 1493 | |
| Human | |
| Supernatant |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto