missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CXCL4/PF4 Antibody, R&D Systems™

Goat Polyclonal Antibody
Marca: R&D Systems AF795
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
CXCL4/PF4 Polyclonal specifically detects CXCL4/PF4 in Human samples. It is validated for Western Blot, Simple Western.
Specifica
| CXCL4/PF4 | |
| Polyclonal | |
| Unconjugated | |
| P02776 | |
| PF4 | |
| E. coli-derived recombinant human CXCL4/PF4 Glu32-Ser101 Accession # P02776 | |
| 100 μg | |
| Primary | |
| Detects human CXCL4/PF4 in direct ELISAs and Western blots. In direct ELISAs, less than 25% cross-reactivity with recombinant mouse CXCL4/PF4 and recombinant rat CXCL4/PF4 is observed, and less than 2% cross-reactivity with recombinant human CXCL7 is observed. | |
| Human | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 1 ug/mL, Simple Western 10 ug/mL | |
| chemokine (C-X-C motif) ligand 4, C-X-C motif chemokine 4, CXCL4iroplact, Iroplact, MGC138298, Oncostatin-A, PF4, PF-4, platelet factor 4, SCYB4oncostatin-A | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 5196 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto