Learn More
Human CXCL9/MIG Antibody, R&D Systems™
Goat Polyclonal Antibody
Marca: R&D Systems AF392-SP
This item is not returnable.
View return policy
Description
CXCL9/MIG Polyclonal specifically detects CXCL9/MIG in Human samples. It is validated for Western Blot, Neutralization, Immunocytochemistry.
Specifications
| CXCL9/MIG | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| chemokine (C-X-C motif) ligand 9, CMK, crg-10, C-X-C motif chemokine 9, Gamma-interferon-induced monokine, Humig, MIG, MIGSmall-inducible cytokine B9, monokine induced by gamma interferon, SCYB9Monokine induced by interferon-gamma | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 4283 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Neutralization, Immunocytochemistry | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Neutralization 3-12 ug/mL, Immunocytochemistry 5-15 ug/mL | |
| Q07325 | |
| CXCL9 | |
| E. coli-derived recombinant human CXCL9/MIG Thr23-Thr125 Accession # Q07325 | |
| 25 μg | |
| Primary | |
| Detects human CXCL9/MIG in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 10% cross-reactivity with recombinant mouse CXCL9 (non-reducing conditions) is observed. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction