Learn More
Human DC-SIGN+DC-SIGNR Antibody, R&D Systems™
Mouse Monoclonal Antibody has been used in 3 publications
Marca: R&D Systems MAB16211-SP
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
DC-SIGN+DC-SIGNR Monoclonal specifically detects DC-SIGN+DC-SIGNR in Human samples. It is validated for Western Blot, Flow Cytometry, Adhesion Blockade, CyTOF-ready.
Specifica
| DC-SIGN+DC-SIGNR | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, Adhesion Blockade 4.0 ug/mL, CyTOF-ready | |
| CD209 | |
| E. coli-derived recombinant human DC-SIGN Extracellular domain | |
| 25 μg | |
| Primary | |
| Detects human DC-SIGN in direct ELISAs and Western blots. Was reported to cross-react with human DC-SIGNR as well as DC-SIGN from Pigtailed Macaque and Rhesus Macaque (10). | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry, Adhesion Blockade, CyTOF | |
| DC28 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 30835 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Titolo del prodotto
For Research Use Only