Learn More
ACROBiosystems Human Fc gamma RIIIA / CD16a (V176) Protein, SUMO,His Tag
287.41€ - 2612.85€
Specifica
Numero di accesso | NP_001121065.1 (F176V) |
---|---|
Peso molecolare | 34.5 kDa |
Sorgente | Human CD16a (V176), SUMO,His Tag (CDA-H5290) is expressed from human 293 cells (HEK293). It contains AA Gly 17 - Gln 208 (Accession # P08637-1 (F176V)). |
Immunogeno | Gly 17 - Gln 208 |
Requisiti di stoccaggio | -20°C |
Codice del prodotto | Marca | Quantità | Prezzo | Quantità e disponibilità | |||||
---|---|---|---|---|---|---|---|---|---|
Codice del prodotto | Marca | Quantità | Prezzo | Quantità e disponibilità | |||||
17196171
|
ACROBiosystems
CDA-H5290-100ug |
100 μg |
365.80€
100µg |
Effettua il login per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso! | |||||
17106181
|
ACROBiosystems
CDA-H5290-1mg |
1 mg |
2612.85€
1mg |
Effettua il login per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso! | |||||
17116181
|
ACROBiosystems
CDA-H5290-50ug |
50 μg |
287.41€
50µg |
Effettua il login per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso! | |||||
Descrizione
CD16 is a low affinity Fc receptor, and has been identified as Fc receptors FcγRIIIa (CD16a) and FcγRIIIb (CD16b). These receptors bind to the Fc portion of IgG antibodies. CD16 encoded by two different highly homologous genes in a cell type-specific manner.CD16 is found on the surface of natural killer cells, neutrophil polymorphonuclear leukocytes, monocytes and macrophages.CD16a antigen is also known as Low affinity immunoglobulin gamma Fc region receptor III-A, Fc-gamma RIII-alpha. CD16b is a low-affinity, GPI-linked receptor expressed by neutrophils and eosinophils, whereas CD16a is an intermediate affinity polypeptide-anchored transmembrane glycoprotein expressed natural killer cells, macrophages, subpopulation of T-cells, immature thymocytes and placentaltrophoblasts.CD16a is involved in phagocytosis, secretion of enzymes and inflammatory mediators, antibodydependent cytotoxicity and clearance of immune complexes. Aberrant expression or mutations of CD16a is implicated in susceptibility to recurrent viral infections, systemic lupus erythematosus, and alloimmune neonatal neutropenia.
This protein carries a SUMO tag at the N-terminus and a polyhistidine tag at the C-terminus. The protein has a calculated MW of 34.5 kDa. The protein migrates as 45-65 kDa under reducing (R) condition (SDS-PAGE) due to Glycosylation.Specifica
NP_001121065.1 (F176V) | |
Human CD16a (V176), SUMO,His Tag (CDA-H5290) is expressed from human 293 cells (HEK293). It contains AA Gly 17 - Gln 208 (Accession # P08637-1 (F176V)). | |
-20°C | |
Fc gamma RIIIA / CD16a | |
Human | |
PBS, pH7.4 | |
HEK293 | |
MABSol™ Fc Receptors |
34.5 kDa | |
Gly 17 - Gln 208 | |
FCGR3A,CD16A,FCG3,FCGR3,IGFR3 | |
Unconjugated | |
His Tag and SUMO Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>90% as determined by SDS-PAGE |