missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Gas6 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 6 publications
Marca: R&D Systems AB885
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Gas6 Polyclonal specifically detects Gas6 in Human samples. It is validated for Western Blot, Blockade of Receptor-ligand Interaction.
Specifica
| Gas6 | |
| Polyclonal | |
| Unconjugated | |
| NP_000811 | |
| GAS6 | |
| Mouse myeloma cell line NS0-derived recombinant human Gas6 Asp118-Ala678 Accession # NP_000811 | |
| 1 mg | |
| Primary | |
| Detects human Gas6 in ELISAs and Western blots. Because this antibody preparation is a total IgG fraction, complete monospecificity cannot be assumed. | |
| Human | |
| Purified |
| Western Blot, Blockade of Receptor-ligand Interaction | |
| LYOPH | |
| Western Blot 1 ug/mL, Blockade of Receptor-ligand Interaction | |
| AXLLG, AXLLGAXL stimulatory factor, AXSFAXL receptor tyrosine kinase ligand, DKFZp666G247, FLJ34709, GAS-6, growth arrest-specific 6, growth arrest-specific protein 6 | |
| Goat | |
| Protein A or G purified | |
| RUO | |
| 2621 | |
| Reconstitute at 1 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto