missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Glutathione S-Transferase pi 1/GSTP1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Marca: R&D Systems MAB6455
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Glutathione S-Transferase pi 1/GSTP1 Monoclonal specifically detects Glutathione S-Transferase pi 1/GSTP1 in Human samples. It is validated for Western Blot, Simple Western, Immunocytochemistry.
Specifica
| Glutathione S-Transferase pi 1/GSTP1 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 0.25 ug/mL, Simple Western 12.5 ug/mL, Immunocytochemistry 8-25 ug/mL | |
| AAC51280 | |
| GSTP1 | |
| E. coli-derived recombinant human Glutathione S-Transferase pi 1/GSTP1 Met1-Glu210 Accession # AAC51280 | |
| 100 μg | |
| Primary | |
| Detects human Glutathione S-Transferase pi 1/GSTP1 in direct ELISAs and Western blots. Detects mouse and rat Glutathione S-Transferase pi 1/GSTP1 in Western Blots. | |
| Human | |
| Purified |
| Western Blot, Immunocytochemistry | |
| 800027 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| deafness, X-linked 7, DFN7, EC 2.5.1.18, FAEES3, fatty acid ethyl ester synthase III, glutathione S-transferase P, glutathione S-transferase pi 1, Glutathione STransferase pi 1, GST class-pi, GST3DFN7, GSTP, GSTP1, GSTP1-1, PI | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 2950 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto