missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Hormone-sensitive Lipase/HSL Antibody, R&D Systems™
Mouse Monoclonal Antibody
Marca: R&D Systems MAB7104
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Hormone-sensitive Lipase/HSL Monoclonal specifically detects Hormone-sensitive Lipase/HSL in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifica
| Hormone-sensitive Lipase/HSL | |
| Monoclonal | |
| LYOPH | |
| Western Blot 2 ug/mL, Immunohistochemistry 8-25 ug/mL | |
| Q05469 | |
| LIPE | |
| E. coli-derived recombinant human Hormone-sensitive Lipase/HSL Met302-Leu425 Accession # Q05469 | |
| 100 μg | |
| Primary | |
| Detects human Hormone-sensitive Lipase/HSL in direct ELISAs. | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry | |
| 729923 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| hormone-sensitive lipase, Hormonesensitive Lipase, hormone-sensitive lipase testicular isoform, HSL, HSLEC 3.1.1.79, LHS, lipase, hormone-sensitive, LIPE | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 3991 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto