missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human IgA ProQuantum Immunoassay Kit

Codice prodotto. p-7173934
Click to view available options
Quantità:
5 x 96 Tests
Dimensione della confezione:
Pezzo
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Codice prodotto. 17184739

Marca: Invitrogen™ A457295

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

ELISA

The ProQuantum Human IgA Immunoassay Kit is designed to provide quantitative measurements of human IgA in small sample volumes. Utilizing proximity-based amplification technology, the assay combines the analyte specificity of high-affinity antibody-antigen binding with the signal detection and amplification capabilities of real-time PCR to achieve a simple yet powerful next-generation protein quantitation platform. A user-friendly workflow combined with intuitive software for analytics enables sample-to-answer in just 2 hours. High sensitivity-detect low levels of protein with greater sensitivity than traditional methods Broad dynamic range-?5 logarithmic units, minimizing sample dilutions to ensure they fall within the range Small sample consumption-use 2-5 μL of sample (compared to 75 μL for triplicate wells with other methods) Fast, easy workflow-2 hours from sample to answer, with no wash steps No proprietary instrument to purchase-runs on any real-time PCR instrument ProQuantum immunoassays utilize a matched pair of target-specific antibodies, each conjugated to a DNA oligonucleotide. During antibody-analyte binding, the two DNA oligos are brought into close proximity, which allows for ligation of the two strands and subsequent creation of a template strand for amplification. This platform leverages the sensitivity and large dynamic range of Applied Biosystems TaqMan real-time PCR technology (Figure 1). The assay workflow...

Human IgA, also known as human immunoglobulin A, is an antibody that is produced by the immune system in response to antigens. It is the most abundant immunoglobulin in mucosal secretions, such as saliva, tears, and breast milk, and plays a key role in defense against pathogens at mucosal surfaces. IgA works by binding to specific antigens and preventing them from entering the body's tissues. It can also activate complement and promote phagocytosis by immune cells. Human IgA can be used in research to study the immune response to pathogens and to develop diagnostic tests and therapeutic interventions for infectious diseases.
TRUSTED_SUSTAINABILITY

Specifica

Numero di accesso P01876, P01877
Intervallo saggio 0.0256-2000 ng/mL
Assay Sensitivity <0.02 ng/mL
Coniugato FAM
Immunoassay Kit Format Proximity-based Immunoassay
Product Type ProQuantum
Tipo di campione Plasma, Serum, Supernatant
Target Species Human
Da utilizzare con (apparecchiatura) qPCR Instruments
Alias gene A2m Marker; Ig alpha 1 Chain C Region; Ig alpha 2 Chain C Region; IgA1; IGHA1; IGHA2; Immunoglobulin A; Immunoglobulin Am1; Immunoglobulin Am2; immunoglobulin heavy constant alpha 1; Immunoglobulin Heavy Constant Alpha 2; immunoglobulin heavy constant alpha 2 (A2m marker)A2m Marker; Ig alpha 1 Chain C Region; Ig alpha 2 Chain C Region; IgA1; IGHA1; IGHA2; Immunoglobulin A; Immunoglobulin Am1; Immunoglobulin Am2; immunoglobulin heavy constant alpha 1; Immunoglobulin Heavy Constant Alpha 2; immunoglobulin heavy constant alpha 2 (A2m marker);
ID gene (immissione) 3493, 3494
Simbolo del gene IGHA1, IGHA2
Contenuto kit Assay Dilution Buffer, Antibody-Conjugate A, Antibody-Conjugate B, Antibody-Conjugate Dilution Buffer, Standard, Master Mix, Ligase
Quantità 5 x 96 Tests
Status giuridico RUO
Shipping Condition Dry Ice
Requisiti di stoccaggio -20°C
Bersaglio Human IgA
Durata del test 30 min
Tempo totale saggio 2 hr
Vedi altri risultati Mostra meno risultati
Correzione del contenuto del prodotto

Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.

Titolo del prodotto

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.

Grazie per averci aiutato a migliorare il nostro sito web. Il vostro feedback è stato inviato