missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IL-17/IL-17A APC-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Marca: R&D Systems IC317A
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
IL-17/IL-17A Monoclonal specifically detects IL-17/IL-17A in Human samples. It is validated for Intracellular Staining by Flow Cytometry.
Specifica
| IL-17/IL-17A | |
| Monoclonal | |
| APC | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| CTLA8, CTLA-8, CTLA8cytotoxic T-lymphocyte-associated serine esterase 8, Cytotoxic T-lymphocyte-associated antigen 8, IL17, IL17A, IL-17A, IL-17Acytotoxic T-lymphocyte-associated protein 8, IL-17CTLA-8, IL17interleukin-17A, interleukin 17 (cytotoxic T-lymphocyte-associated serine esterase 8), interleukin 17A | |
| Mouse | |
| Protein A or G purified from ascites | |
| RUO | |
| 3605 | |
| Human | |
| Purified |
| Flow Cytometry | |
| 41809 | |
| Intracellular Staining by Flow Cytometry 10 uL/10^6 cells | |
| Q16552 | |
| IL17A | |
| E. coli-derived recombinant human IL-17/IL-17A Ile20-Ala155 Accession # Q16552 | |
| 100 Tests | |
| Primary | |
| Detects human IL-17/IL-17A in direct ELISAs. In direct ELISAs, approximately 12% cross-reactivity with recombinant canine IL-17/IL-17A is observed and 25%-50% reactivity with recombinant human (rh) IL-17A/IL-17F heterodimer is observed. No cross-reactivity with recombinant mouse IL-17/IL-17A, rhIL-17B, rhIL-17C, rhIL-17D, rhIL-17E, or rhIL-17F is observed. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
| IgG2b |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto