Learn More
R&D Systems™ Human IL-17 Quantikine ELISA Kit
Codice prodotto. 18772304
Click to view available options
Quantità:
1 Plate
6 Plates
Dimensione della confezione:
1 set
Codice prodotto. 18772304
Marca: R&D Systems™ D1700
Human IL-17 Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.
The Human IL-17 Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human IL-17/IL-17A in cell culture supernates (100 ul), serum (100 ul), edta plasma (100 ul), heparin plasma (100 ul), citrate plasma (100 ul).
TRUSTED_SUSTAINABILITY
Specifica
Intervallo saggio 31.20 - 2000pg/mL
Assay Sensitivity 15pg/mL
Coniugato HRP
Cross-reattività <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
Product Type Solid Phase Sandwich ELISA
Tipo di campione Cell Culture Supernatant, Plasma (EDTA, Citrate, Heparin), Serum
Specificità Natural and Recombinant Human IL-17
Sufficiente per 1 x 96-well Plate
Target Species Human
Da utilizzare con (applicazione) ELISA
ID gene (immissione) 3605
Quantità 1 Plate
Volume del campione Cell Culture Supernates (100uL),Serum (100uL),EDTA Plasma (100uL),Heparin Plasma (100uL),Citrate Plasma (100uL)
Requisiti di stoccaggio Store the unopened product at 2° to 8°C. Do not use past expiration date.
Sinonimo CTLA8, CTLA-8, CTLA8cytotoxic T-lymphocyte-associated serine esterase 8, Cytotoxic T-lymphocyte-associated antigen 8, IL17, IL17A, IL-17 A, IL-17 Acytotoxic T-lymphocyte-associated protein 8, IL-17 CTLA-8, IL17interleukin-17 A, interleukin 17 (cytotoxic T-lymphocyte-associated serine esterase 8), interleukin 17 A