missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IL-1ra/IL-1F3 Biotinylated Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 5 publications
Marca: R&D Systems BAF280
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
IL-1ra/IL-1F3/IL1RN Polyclonal specifically detects IL-1ra/IL-1F3/IL1RN in Human samples. It is validated for Western Blot, ELISA Detection (Matched Antibody Pair).
Specifica
| IL-1ra/IL-1F3/IL1RN | |
| Polyclonal | |
| Biotin | |
| DIRA, ICIL-1RA, IL1F3, IL-1F3, IL-1ra, IL1RA, IL-1ra3, IL1RN, IL-1RN, interleukin 1 receptor antagonist, IRAP, MVCD4 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 3557 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, ELISA | |
| LYOPH | |
| Western Blot 0.1 ug/mL, ELISA Detection (Matched Antibody Pair) 0.1-0.4 ug/mL | |
| IL1RN | |
| E. coli-derived recombinant human IL-1ra/IL-1F3 | |
| 50 μg | |
| Primary | |
| Detects human IL-1ra/IL-1F3 in ELISAs and Western blots. In sandwich immunoassays, less than 0.2% cross-reactivity with recombinant mouse IL-1ra, recombinant human (rh) IL-1 alpha , rhIL-1 beta , rhIL-1 RI, and rhIL-1 RII is observed. | |
| Human | |
| IgG |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto