Learn More
R&D Systems™ Human IL-33 DuoSet ELISA
Codice prodotto. p-7079219
Click to view available options
Quantità:
1 Kit (for 15 Plates)
1 Kit (for 5 Plates)
Dimensione della confezione:
1 set
pezzi 15
Codice prodotto. 30020997
Marca: R&D Systems™ DY3625B
Includes
Capture antibody, detection antibody, recombinant standard, streptavidin conjugated to horseradish-peroxidase (Streptavidin-HRP) and development protocols
Human IL-33 DuoSet ELISA from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.
The Human IL-33 DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies human IL-33.
TRUSTED_SUSTAINABILITY
Specifica
Descrizione Fucose Detection
Product Type ELISA Kit
Quantità 1 Kit (for 15 Plates)
Specie Human
Intervallo saggio 23.40 to 1500 pg/mL
Cross-reattività Not observed
Formato 96-well Strip Plate
Da utilizzare con (applicazione) ELISA
Tipo di campione Cell Culture Supernatant, Serum, Plasma
Contenuto kit Capture antibody, Detection antibody, Recombinant standard, Streptavidin conjugated to horseradish-peroxidase (Streptavidin-HRP) and development protocols
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.