Learn More
Novus Biologicals™ Human IL-4R alpha (Research Grade Dupilumab Biosimilar) Antibody, Novus Biologicals™
Human Monoclonal Antibody
Marca: Novus Biologicals™ MAB11481-100
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
IL-4R alpha Monoclonal antibody specifically detects IL-4R alpha in Human samples. It is validated for ELISA
Specifica
| IL-4R alpha | |
| Monoclonal | |
| Unconjugated | |
| P24394 | |
| Human | |
| Protein A or G purified from ascites | |
| RUO | |
| 3566 | |
| Human | |
| Purified |
| ELISA | |
| Hu209 | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. See Certificate of Analysis for details. *Small pack size (-SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
| CD124, CD124 antigen, IL-4 R alpha, IL-4 receptor subunit alpha, IL4R, IL4R alpha, IL-4R subunit alpha, IL-4RA, IL4RACD124, IL-4R-alpha, interleukin 4 receptor, interleukin-4 receptor alpha chain, interleukin-4 receptor subunit alpha | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human IL-4 R, Gly24-His232, Accession # P24394 | |
| 100 μg | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. For liquid material, refer to CoA for concentration. | |
| Use a manual defrost freezer and avoid repeated freeze/thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 6 months, -20°C to -70°C under sterile conditions after reconstitution. | |
| IgG1 |