missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IQGAP1 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Marca: R&D Systems AF5069-SP
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
IQGAP1 Polyclonal specifically detects IQGAP1 in Human samples. It is validated for Western Blot.
Specifica
| IQGAP1 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| HUMORFA01, IQ motif containing GTPase activating protein 1, KIAA0051ras GTPase-activating-like protein IQGAP1, p195, p195RasGAP-like with IQ motifs, SAR1 | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 8826 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| P46940 | |
| IQGAP1 | |
| E. coli-derived recombinant human IQGAP1 Lys505-Ser717 Accession # P46940 | |
| 25 μg | |
| Primary | |
| Detects endogenous human IQGAP1 in Western blots. | |
| Human | |
| IgG |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
For Research Use Only
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto