Learn More
Human KCC2/SLC12A5 Antibody, R&D Systems™
GREENER_CHOICE
Mouse Monoclonal Antibody
Marca:  R&D Systems MAB8369-SP
Codice prodotto. 18702819
  • ONLINE EXCLUSIVE PRICE
    251.00€
    237.30€ / 25µg
    Risparmia 13.70€
    5% Sconto
Questo articolo non è restituibile. Consulta la politica di reso
Descrizione
Descrizione
KCC2/SLC12A5 Monoclonal specifically detects KCC2/SLC12A5 in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.
TRUSTED_SUSTAINABILITY
Specifica
Specifica
KCC2/SLC12A5
Monoclonal
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Electroneutral potassium-chloride cotransporter 2, hKCC2, KCC2K-Cl cotransporter 2, KIAA1176erythroid K-Cl cotransporter 2, Neuronal K-Cl cotransporter, SLC12A5, solute carrier family 12 (potassium/chloride transporter), member 5, solute carrier family 12 member 5
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
57468
Reconstitute at 0.5 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
IgG1
Western Blot, Flow Cytometry, CyTOF
905705
Western Blot 0.5 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready
Q9H2X9
SLC12A5
NS0 mouse myeloma cell line transfected with human SLC12A5 Accession # Q9H2X9
25 μg
Primary
Detects human SLC12A5 in ELISAs and Western blots.
Human
Purified
Suggerimenti di prodotto
Suggerimenti di prodotto
Videos
SDS
Documenti
Documenti
Certificati
Promozioni
Promozioni