Learn More
ACROBiosystems Human latent GDF-8 Protein, His Tag
Marca: ACROBiosystems GD8-H5243-500ug
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
Growth differentiation factor 8 (GDF8), also known as myostatin, is a unique member of the transforming growth factor-β superfamily that is expressed in human granulosa cells and has important roles in regulating a variety of ovarian functions. GDF8 acts as a negative regulator of skeletal muscle growth and differentiation. In addition to the expression in the musculoskeletal system, GDF8 is also expressed in various tissues, including the reproductive system.
This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 42 kDa.Specifica
42 kDa | |
Human latent GDF-8, His Tag (GD8-H5243) is expressed from human 293 cells (HEK293). It contains AA Asn 24 - Ser 375 (Accession # O14793-1). | |
-20°C | |
latent GDF-8 | |
Human | |
HEK293 | |
Bio-Markers & CD Antigens |
500 μg | |
Asn 24 - Ser 375 | |
latent GDF-8, GDF-8, Myostatin, GDF8, MSTN, Growth/differentiation factor 8 | |
Unconjugated | |
His Tag | |
Powder |