Learn More
R&D Systems™ Human MBP DuoSet ELISA
Click to view available options
Quantità:
1 Kit (for 5 Plates)
Dimensione della confezione:
pezzi 5
Descrizione
Includes
Capture antibody, detection antibody, recombinant standard, streptavidin conjugated to horseradish-peroxidase (Streptavidin-HRP) and development protocols
The Human MBP DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies human MBP.
Specifica
Specifica
| Quantità | 1 Kit (for 5 Plates) |
| Specie | Human |
| Coniugato | Biotin |
| Bersaglio legante | MBP |
| Intervallo saggio | 234.40 to 15000 pg/mL |
| Da utilizzare con (applicazione) | ELISA |
| ID gene (immissione) | 4155 |
| Sinonimo | Hmbpr, MGC99675, mld, Myelin A1 protein, myelin basic protein, Myelin membrane encephalitogenic protein, shi |
| Content And Storage | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |