missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse S100A4 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Marca: R&D Systems AF4138-SP
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
S100A4 Polyclonal specifically detects S100A4 in Human, Mouse samples. It is validated for Western Blot, Simple Western, Knockout Validated.
Specifica
| S100A4 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| 18A2, 42A, Calvasculin, CAPL, CAPLS100 calcium binding protein A4 (calcium protein, calvasculin, metastasin, fibroblast-specific protein-1,42A, FSP1, leukemia multidrug resistance associated protein, malignant transformation suppression 1, Metastasin, MTS1, murine placental homolog), P9KA, PEL98, Placental calcium-binding protein, Protein Mts1, protein S100-A4, S100 calcium binding protein A4, S100 calcium-binding protein A4, S100 calcium-binding protein A4 (calcium protein, calvasculin, metastasin, murine placental homolog) | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 6275 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, KnockDown | |
| LYOPH | |
| Western Blot 1 ug/mL, Simple Western 10 ug/mL, Knockout Validated | |
| P07091 | |
| S100A4 | |
| E. coli-derived recombinant mouse S100A4 Ala2-Lys101 Accession # P07091 | |
| 25 μg | |
| Primary | |
| Detects human and mouse S100A4 in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant mouse (rm) S100A8, rmS100A6, rmS100A9, rmS100A10, rmS100A13, rmS100A16, recombinant human (rh) S100A1, rhS100A7, rhS100A11, rhS100B, and rhS100P is observed. | |
| Human, Mouse | |
| IgG |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
For Research Use Only
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto