missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human P-Cadherin APC-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Marca: R&D Systems FAB861A
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
P-Cadherin Monoclonal specifically detects P-Cadherin in Human samples. It is validated for Flow Cytometry, Knockout Validated.
Specifica
| P-Cadherin | |
| Monoclonal | |
| APC | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| CAD3, cadherin 3, P-cadherin (placental), cadherin 3, type 1, P-cadherin (placental), cadherin-3, CDH3, CDHP, CDHPcalcium-dependent adhesion protein, placental, HJMD, PCAD, PCadherin, PCADP-cadherin, Placental cadherin | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 1001 | |
| Human | |
| Purified |
| Flow Cytometry, KnockDown | |
| 104805 | |
| Flow Cytometry 10 uL/10^6 cells, Knockout Validated 10 uL/10^6 cells | |
| CAA45177 | |
| CDH3 | |
| Mouse myeloma cell line NS0-derived recombinant human P-Cadherin Asp108-Gly654 Accession # CAA45177 | |
| 100 Tests | |
| Primary | |
| Detects human P-Cadherin in direct ELISAs and Western blots. In Western blots, does not cross-react with recombinant human (rh) Cadherin-8, recombinant mouse P-Cadherin, or rhVE-Cadherin. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
| IgG1 |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto