missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human p16INK4a/ CDKN2A Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 5 publications
Marca: R&D Systems AF5779
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
p16INK4a/CDKN2A Polyclonal specifically detects p16INK4a/CDKN2A in Human samples. It is validated for Western Blot, Simple Western, Immunocytochemistry.
Specifica
| p16INK4a/CDKN2A | |
| Polyclonal | |
| Unconjugated | |
| P42771 | |
| CDKN2A | |
| E. coli-derived recombinant human p16INK4a/CDKN2A Glu2-Asp156 Accession # P42771 | |
| 100 μg | |
| Primary | |
| Detects human p16INK4a/CDKN2A in Western blots. | |
| Human | |
| IgG |
| Western Blot, Immunocytochemistry | |
| LYOPH | |
| Western Blot 1 ug/mL, Simple Western 10 ug/mL, Immunocytochemistry 0.3-15 ug/mL | |
| ARF, CDK4 inhibitor p16-INK4, CDK4I, CDK4IP14ARF, CDKN2, CDKN2A, cell cycle negative regulator beta, CMM2P16-INK4A, Cyclin-dependent kinase 4 inhibitor A, cyclin-dependent kinase inhibitor 2A, cyclin-dependent kinase inhibitor 2A (melanoma, p16, inhibits CDK4), INK4, INK4A, MLM, MLMP16INK4, MTS-1, MTS1P14, Multiple tumor suppressor 1, p14, p14ARF, p16, p16-INK4, p16INK4a, p16-INK4a, P19, p19ARF, TP16 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 1029 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto