missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human PDGF-AA Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 6 publications
Marca: R&D Systems AF-221-NA
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
PDGF-AA Polyclonal specifically detects PDGF-AA in Human samples. It is validated for Western Blot, Immunohistochemistry, Neutralization.
Specifica
| PDGF-AA | |
| Polyclonal | |
| Unconjugated | |
| P04085 | |
| PDGFA | |
| E. coli-derived recombinant human PDGF-AA Ser87-Thr211 Accession # P04085 | |
| 100 μg | |
| Primary | |
| Detects human PDGF-AA and PDGF-AB in direct ELISAs and Western blots. In Western blots, less than 5% cross-reactivity with recombinant human PDGF-BB and recombinant rat PDGF-BB is observed. | |
| Human | |
| IgG |
| Western Blot, Immunohistochemistry, Neutralization | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL, Neutralization 0.1-0.3 ug/mL | |
| PDGF1, PDGF-A, PDGFAA, PDGFAB | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 5154 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto