missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human PDGF-BB Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 14 publications
Marca: R&D Systems AF-220-NA
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
PDGF-BB Polyclonal specifically detects PDGF-BB in Human samples. It is validated for Western Blot, Neutralization.
Specifica
| PDGF-BB | |
| Polyclonal | |
| Unconjugated | |
| P01127 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 5155 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Neutralization | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Neutralization 0.1-0.5 ug/mL | |
| PDGFB | |
| E. coli-derived recombinant human PDGF-BB Ser82-Thr190 Accession # P01127 | |
| 100 μg | |
| Primary | |
| Detects PDGF-BB in direct ELISAs and Western blots. | |
| Human | |
| IgG |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto